missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Manufacturer:  Alfa Aesar™ H63920MD

 View more versions of this product

Catalog No. AAH63920MD



Specifications

Specifications

Yellow to Brown
6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
C13H13FN2
8627107
FBWKDFSEFVFKFY-UHFFFAOYSA-N
6-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
11053072
Solid
250mg
180161-14-2
MFCD04114772
4-6-fluoro-1h-indol-3-yl-1236-tetrahydropyridine, 3-1236-tetrahydropyridine-4-yl-6-fluoro-1h-indole, 6-fluoro-3-1236-tetrahydro-pyridin-4-yl-1h-indole, 6-fluoro-3-1236-tetrahydropyridin-4-yl-1h-indole, 6-fluoro-3-1236-tetrahydro-4-pyridinyl-1h-indole, 6-fluoro-3-1236-tetrahydro4-pyridinyl-1h-indole, 4-6-fluoroindol-3-yl-1256-tetrahydropyridine, 6-fluoro-3-4-1256-tetrahydropyridyl indole
C1CNCC=C1C2=CNC3=C2C=CC(=C3)F
216.259
216.25
97%

Chemical Identifiers

180161-14-2
216.259
FBWKDFSEFVFKFY-UHFFFAOYSA-N
11053072
C1CNCC=C1C2=CNC3=C2C=CC(=C3)F
C13H13FN2
MFCD04114772
4-6-fluoro-1h-indol-3-yl-1236-tetrahydropyridine, 3-1236-tetrahydropyridine-4-yl-6-fluoro-1h-indole, 6-fluoro-3-1236-tetrahydro-pyridin-4-yl-1h-indole, 6-fluoro-3-1236-tetrahydropyridin-4-yl-1h-indole, 6-fluoro-3-1236-tetrahydro-4-pyridinyl-1h-indole, 6-fluoro-3-1236-tetrahydro4-pyridinyl-1h-indole, 4-6-fluoroindol-3-yl-1256-tetrahydropyridine, 6-fluoro-3-4-1256-tetrahydropyridyl indole
6-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents