missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluoro-2-methyl-3-nitropyridine, 98%

$127.77 - $509.49

Chemical Identifiers

CAS 18605-16-8
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD03095071
InChI Key JVLRFRZVCOWBDY-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitro-6-methyl pyridine, 2-fluoro-5-nitro-6-methylpyridin, 2-fluoro-5-nitro-6-methylpyridine, 2-fluoro-5-nitro-6-picoline, 2-fluoro-6-methyl-5-nitropyridine, 6-fluoro-3-nitro-2-picoline, acmc-209enj, pubchem6327, pyridine, 6-fluoro-2-methyl-3-nitro, pyridine,6-fluoro-2-methyl-3-nitro
PubChem CID 12408185
IUPAC Name 6-fluoro-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(=N1)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6187706
View Documents
Alfa Aesar™
H6187706
5g Each for $127.77
 
AAH6187714
View Documents
Alfa Aesar™
H6187714
25g Each for $509.49
 
Specifications

Chemical Identifiers

18605-16-8
156.116
JVLRFRZVCOWBDY-UHFFFAOYSA-N
12408185
CC1=C(C=CC(=N1)F)[N+](=O)[O-]
C6H5FN2O2
MFCD03095071
2-fluoro-5-nitro-6-methyl pyridine, 2-fluoro-5-nitro-6-methylpyridin, 2-fluoro-5-nitro-6-methylpyridine, 2-fluoro-5-nitro-6-picoline, 2-fluoro-6-methyl-5-nitropyridine, 6-fluoro-3-nitro-2-picoline, acmc-209enj, pubchem6327, pyridine, 6-fluoro-2-methyl-3-nitro, pyridine,6-fluoro-2-methyl-3-nitro
6-fluoro-2-methyl-3-nitropyridine

Specifications

1.5216
18605-16-8
MFCD03095071
JVLRFRZVCOWBDY-UHFFFAOYSA-N
6-fluoro-2-methyl-3-nitropyridine
12408185
Solid
6-Fluoro-2-methyl-3-nitropyridine
5g
C6H5FN2O2
2-fluoro-5-nitro-6-methyl pyridine, 2-fluoro-5-nitro-6-methylpyridin, 2-fluoro-5-nitro-6-methylpyridine, 2-fluoro-5-nitro-6-picoline, 2-fluoro-6-methyl-5-nitropyridine, 6-fluoro-3-nitro-2-picoline, acmc-209enj, pubchem6327, pyridine, 6-fluoro-2-methyl-3-nitro, pyridine,6-fluoro-2-methyl-3-nitro
CC1=C(C=CC(=N1)F)[N+](=O)[O-]
156.116
156.11
98%
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures