Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Fluoro-2,3-dimethoxybenzoic acid, 95%

Chemical Identifiers

Chemical Name or Material 6-Fluoro-2,3-dimethoxybenzoic acid
CAS 265670-72-2
Formula Weight 200.17
MDL Number MFCD13193706
Beilstein 8482222
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66209MD

 
alfa aesar™
H66209MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.17
Chemical Name or Material 6-Fluoro-2,3-dimethoxybenzoic acid
Physical Form Solid
MDL Number MFCD13193706
CAS 265670-72-2
Beilstein 8482222
Quantity 250mg
Assay 95%