Please login to your online account to display your discounted pricing

6-Fluoro-2,3-dihydro-4H-thiochromen-4-one, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Fluoro-2, 3-dihydro-4H-thiochromen-4-one
CAS 21243-18-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7FOS
Formula Weight 182.22
MDL Number MFCD00078488
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW05178DA

  SDS
fisher scientific company
SEW05178DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SEW05178EA

  SDS
fisher scientific company
SEW05178EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.22
Quantity 1g
Assay 95%
CAS Min % 95.0
CAS 21243-18-5
Chemical Name or Material 6-Fluoro-2, 3-dihydro-4H-thiochromen-4-one
MDL Number MFCD00078488
Packaging Amber glass bottle
Assay Percent Range 95%
Molecular Formula C9H7FOS
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.