missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Fluoro-1-indanone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442330050

 View more versions of this product

Catalog No. AC442330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

6-Fluoro-1-indanone
White to Brown
5g
96.0
96% min. (GC)
1h-inden-1-one, 6-fluoro-2,3-dihydro, 6-fluoro-1-indanone, 6-fluoro-2,3-dihydro-1h-inden-1-one, 6-fluoro-indan-1-one, 6-fluoroindan-1-one, 6-fluoroindanone, 6-fluroro-1-indanone, 6fluoro-1-indanone, acmc-1bwfy, pubchem8856
C1CC(=O)C2=C1C=CC(=C2)F
150.15
150.15
97%
Authentic
58°C to 61°C
1481-32-9
100.0
C9H7FO
LVUUCFIQQHEFEJ-UHFFFAOYSA-N
6-fluoro-2,3-dihydroinden-1-one
1519464
Crystalline Powder

Chemical Identifiers

1481-32-9
150.15
1h-inden-1-one, 6-fluoro-2,3-dihydro, 6-fluoro-1-indanone, 6-fluoro-2,3-dihydro-1h-inden-1-one, 6-fluoro-indan-1-one, 6-fluoroindan-1-one, 6-fluoroindanone, 6-fluroro-1-indanone, 6fluoro-1-indanone, acmc-1bwfy, pubchem8856
6-fluoro-2,3-dihydroinden-1-one
C9H7FO
LVUUCFIQQHEFEJ-UHFFFAOYSA-N
1519464
C1CC(=O)C2=C1C=CC(=C2)F
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents