missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Ethoxypyridine-3-boronic acid, 98%

$63.11 - $621.86

Chemical Identifiers

CAS 612845-44-0
Molecular Formula C7H10BNO3
Formula Weight 166.97
MDL Number MFCD06201032
Beilstein 9388808
Synonym 2-Ethoxy-5-pyridylboronic acid
Chemical Name or Material 6-Ethoxypyridine-3-boronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2039803 SDS Alfa Aesar™
L2039803
1g Each for $166.24
AAL2039806 SDS Alfa Aesar™
L2039806
5g Each for $621.86
AAL20398MD SDS Alfa Aesar™
L20398MD
250mg Each for $63.11
Specifications Description & Specifications

Chemical Identifiers

612845-44-0
166.97
9388808
6-Ethoxypyridine-3-boronic acid
C7H10BNO3
MFCD06201032
2-Ethoxy-5-pyridylboronic acid

Specifications

102° to 104°C
612845-44-0
166.97
9388808
6-Ethoxypyridine-3-boronic acid
1g
C7H10BNO3
MFCD06201032
2-Ethoxy-5-pyridylboronic acid
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning