missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Ethoxy-2-naphthaleneboronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440230050
Specifications
6-Ethoxy-2-naphthaleneboronic acid | |
White | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-ethoxy-6-naphthaleneboronic acid, 6-ethoxy-2, 6-ethoxy-2-naphthaleneboronic acid, 6-ethoxy-2-naphthalenyl-boronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, acmc-209ies, boronic acid, 6-ethoxy-2-naphthalenyl, boronic acid, b-6-ethoxy-2-naphthalenyl, boronic acid,b-6-ethoxy-2-naphthalenyl | |
B(C1=CC2=C(C=C1)C=C(C=C2)OCC)(O)O | |
216.04 | |
216.04 | |
97% |
Authentic | |
238.5°C to 243.3°C | |
352525-98-5 | |
100.0 | |
C12H13BO3 | |
INXXVGFSXYJGHI-UHFFFAOYSA-N | |
(6-ethoxynaphthalen-2-yl)boronic acid | |
4641693 | |
Powder |
Chemical Identifiers
352525-98-5 | |
216.04 | |
2-ethoxy-6-naphthaleneboronic acid, 6-ethoxy-2, 6-ethoxy-2-naphthaleneboronic acid, 6-ethoxy-2-naphthalenyl-boronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, acmc-209ies, boronic acid, 6-ethoxy-2-naphthalenyl, boronic acid, b-6-ethoxy-2-naphthalenyl, boronic acid,b-6-ethoxy-2-naphthalenyl | |
(6-ethoxynaphthalen-2-yl)boronic acid |
C12H13BO3 | |
INXXVGFSXYJGHI-UHFFFAOYSA-N | |
4641693 | |
B(C1=CC2=C(C=C1)C=C(C=C2)OCC)(O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning