Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(Diethoxymethyl)uracil, 98%

$305.04 - $305.04

Chemical Identifiers

Chemical Name or Material 6-(Diethoxymethyl)uracil
CAS 16953-48-3
Molecular Formula C9H14N2O4
Formula Weight 214.22g/mol
MDL Number MFCD12755731
Synonym 6-(Diethoxymethyl)pyrimidine-2,4(1H,3H)-dione; Uracil-6-carboxaldehyde diethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5169403

 
alfa aesar™
H51694-03
Each for $305.04
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 6-(Diethoxymethyl)uracil
CAS 16953-48-3
Assay 98%
Molecular Formula C9H14N2O4
Formula Weight 214.22g/mol
MDL Number MFCD12755731
Synonym 6-(Diethoxymethyl)pyrimidine-2,4(1H,3H)-dione; Uracil-6-carboxaldehyde diethyl acetal