missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H50139MD

 View more versions of this product

Catalog No. AAH50139MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester
C15H22BNO3
250 mg
2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3
275.155
275.16
947191-69-7
MFCD07781184
Air Sensitive
QEQNYJUPYICBCR-UHFFFAOYSA-N
2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
24208797
97%

Chemical Identifiers

947191-69-7
275.155
QEQNYJUPYICBCR-UHFFFAOYSA-N
24208797
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3
C15H22BNO3
MFCD07781184
2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only