Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-[Cyclohexyl(methyl)amino]pyridine-3-boronic acid pinacol ester, 95%

$344.93 - $344.93

Chemical Identifiers

Chemical Name or Material 6-[Cyclohexyl(methyl)amino]pyridine-3-boronic acid pinacol ester
CAS 1073354-73-0
Molecular Formula C18H29BN2O2
Formula Weight 316.25g/mol
MDL Number MFCD09027071
Synonym N-Cyclohexyl-N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2701303

 
alfa aesar™
H27013-03
1g Each for $344.93
Description & Specifications

Specifications

Melting Point 107° to 110°C
Quantity 1g
Chemical Name or Material 6-[Cyclohexyl(methyl)amino]pyridine-3-boronic acid pinacol ester
CAS 1073354-73-0
Assay 95%
Molecular Formula C18H29BN2O2
Formula Weight 316.25g/mol
MDL Number MFCD09027071
Synonym N-Cyclohexyl-N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.