Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Cyanonicotinic acid, 97%

$118.84 - $118.84

Chemical Identifiers

Chemical Name or Material 6-Cyanonicotinic acid
CAS 70165-31-0
Molecular Formula C7H4N2O2
Formula Weight 148.12
MDL Number MFCD01318135
Beilstein 4384027
Synonym 5-Carboxypicolinonitrile; 2-Cyano-5-carboxypyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL17127MD

  SDS
alfa aesar™
L17127MD
250mg Each for $118.84
Description & Specifications

Specifications

Formula Weight 148.12
Assay 97%
Molecular Formula C7H4N2O2
CAS 70165-31-0
Synonym 5-Carboxypicolinonitrile; 2-Cyano-5-carboxypyridine
Health Hazard 1 Danger
Chemical Name or Material 6-Cyanonicotinic acid
MDL Number MFCD01318135
Beilstein 4384027
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 189° to 194°C
Quantity 250mg

Applications
6-Cyanonicotinic acid, is used in medicine

Solubility
Soluble in alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.