missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Cyanoindole 98.0+%, TCI America™
Supplier: TCI America C208425G
Specifications
| 6-Cyanoindole | |
| 130°C | |
| MFCD00016732 | |
| 3439 | |
| SZSZDBFJCQKTRG-UHFFFAOYSA-N | |
| 1H-indole-6-carbonitrile | |
| 85146 | |
| ≥98.0% (GC) |
| 15861-36-6 | |
| C9H6N2 | |
| 25 g | |
| 6-cyanoindole, 6-indolecarbonitrile, indole-6-carbonitrile, 6-cyano-1h-indole, indole-6-cyano, 6-cyano-indole, 6-indole carbonitrile, pubchem7239, cyanoindole-6, 1h-indol-6-yl cyanide | |
| C1=CC(=CC2=C1C=CN2)C#N | |
| 142.161 | |
| 142.16 | |
| Crystalline Powder |
Chemical Identifiers
| 15861-36-6 | |
| 142.161 | |
| SZSZDBFJCQKTRG-UHFFFAOYSA-N | |
| 85146 | |
| C1=CC(=CC2=C1C=CN2)C#N |
| C9H6N2 | |
| MFCD00016732 | |
| 6-cyanoindole, 6-indolecarbonitrile, indole-6-carbonitrile, 6-cyano-1h-indole, indole-6-cyano, 6-cyano-indole, 6-indole carbonitrile, pubchem7239, cyanoindole-6, 1h-indol-6-yl cyanide | |
| 1H-indole-6-carbonitrile |
Safety and Handling
TSCA : No