missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Cyano-2-(2-pyridyl)benzimidazole, 97%

Manufacturer:  Alfa Aesar™ H5861503

 View more versions of this product

Catalog No. AAH5861503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Cyano-2-(2-pyridyl)benzimidazole
1g
C13H8N4
UN3439
JOPNJLSDCGCEFK-UHFFFAOYSA-N
2-pyridin-2-yl-3H-benzimidazole-5-carbonitrile
11543088
Solid
Brown
226702-84-7
MFCD22581244
2-pyridin-2-yl-1h-1,3-benzodiazole-5-carbonitrile, 2-pyridin-2-yl-3h-1,3-benzodiazole-5-carbonitrile, 6-cyano-2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzimidazole-5-carbonitrile, 2-pyridin-2-yl-1h-benzoimidazole-5-carbonitrile, 2-pyridin-2-yl-1h-benzo d imidazole-5-carbonitrile
C1=CC=NC(=C1)C2=NC3=C(N2)C=C(C=C3)C#N
220.235
220.23
97%

Chemical Identifiers

226702-84-7
220.235
JOPNJLSDCGCEFK-UHFFFAOYSA-N
11543088
C1=CC=NC(=C1)C2=NC3=C(N2)C=C(C=C3)C#N
C13H8N4
MFCD22581244
2-pyridin-2-yl-1h-1,3-benzodiazole-5-carbonitrile, 2-pyridin-2-yl-3h-1,3-benzodiazole-5-carbonitrile, 6-cyano-2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzimidazole-5-carbonitrile, 2-pyridin-2-yl-1h-benzoimidazole-5-carbonitrile, 2-pyridin-2-yl-1h-benzo d imidazole-5-carbonitrile
2-pyridin-2-yl-3H-benzimidazole-5-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents