missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Cyano-1-methylindole, 97%

Manufacturer:  Alfa Aesar™ H6385306

 View more versions of this product

Catalog No. AAH6385306



Specifications

Specifications

6-Cyano-1-methylindole
5g
C10H8N2
UN3439
1-methyl-1h-indole-6-carbonitrile, 1h-indole-6-carbonitrile, 1-methyl, 6-cyano-1-methylindole, acmc-1cirg, 1-methyl-6-cyanoindole, 6-cyano-1-methyl-1h-indole, 1h-indole-6-carbonitrile, 1-methyl-9ci
CN1C=CC2=C1C=C(C=C2)C#N
156.188
156.18
97%
White to Yellow
20996-87-6
MFCD08690254
472731
FOGAAHLMYPQCJG-UHFFFAOYSA-N
1-methylindole-6-carbonitrile
15384493
Solid

Chemical Identifiers

20996-87-6
156.188
FOGAAHLMYPQCJG-UHFFFAOYSA-N
15384493
CN1C=CC2=C1C=C(C=C2)C#N
C10H8N2
MFCD08690254
1-methyl-1h-indole-6-carbonitrile, 1h-indole-6-carbonitrile, 1-methyl, 6-cyano-1-methylindole, acmc-1cirg, 1-methyl-6-cyanoindole, 6-cyano-1-methyl-1h-indole, 1h-indole-6-carbonitrile, 1-methyl-9ci
1-methylindole-6-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents