missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloroquinoline, TCI America™

$77.92 - $218.48

Chemical Identifiers

CAS 612-57-7
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00024024
InChI Key GKJSZXGYFJBYRQ-UHFFFAOYSA-N
Synonym quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-,
PubChem CID 69163
IUPAC Name 6-chloroquinoline
SMILES C1=CC2=C(C=CC(=C2)Cl)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C02831G View Documents TCI America
C02831G
1g Each for $77.92
 
C02835G View Documents TCI America
C02835G
5g Each for $218.48
 
Specifications

Chemical Identifiers

612-57-7
163.604
GKJSZXGYFJBYRQ-UHFFFAOYSA-N
69163
C1=CC2=C(C=CC(=C2)Cl)N=C1
C9H6ClN
MFCD00024024
quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-,
6-chloroquinoline

Specifications

264°C
41°C
612-57-7
MFCD00024024
GKJSZXGYFJBYRQ-UHFFFAOYSA-N
6-chloroquinoline
69163
Crystal-Lump at 20°C
6-Chloroquinoline
Undesignated
1g
C9H6ClN
quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-,
C1=CC2=C(C=CC(=C2)Cl)N=C1
163.604
163.60
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator