Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloroquinoline, 99%

$57.57 - $204.92

Chemical Identifiers

Chemical Name or Material 6-Chloroquinoline
CAS 612-57-7
Molecular Formula C9H6ClN
Formula Weight 163.61
MDL Number MFCD00024024
Beilstein 112963
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2016403

  SDS
alfa aesar™
L2016403
1g Each for $57.57

AAL2016406

  SDS
alfa aesar™
L2016406
5g Each for $204.92
Description & Specifications

Specifications

Formula Weight 163.61
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 99%
MDL Number MFCD00024024
Health Hazard 1 Warning
Flash Point >110°C (230°F)
Chemical Name or Material 6-Chloroquinoline
Beilstein 112963
CAS 612-57-7
Molecular Formula C9H6ClN
Boiling Point 264°C
Quantity 1g
TSCA TSCA
Odor Characteristic
Melting Point 28° to 42°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
6-Chloroquinoline may be used in the synthesis of 3-methyl-3H-imidazo[4,5-f]quinolin-2-amine. It may be used as catalyst for high yield preparation of ethoxycarbonyl isothiocyanate. Catalyst for high yield preparation of ethoxycarbonyl isothiocyanate. The standard molar enthalpy of formation of 6-chloroquinoline has been derived from the standard molar enthalpy of combustion and was evaluated in terms of molecular structure.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.