Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloropyridine-3-boronic acid, 96%


Quantity 250mg

Manufacturer: alfa aesar™  L20388MD

 View more versions of this product

Catalog No. AAL20388MD

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 157.37
Chemical Name or Material 6-Chloropyridine-3-boronic acid
MDL Number MFCD03094998
Assay 96%
Molecular Formula C5H5BClNO2
CAS 444120-91-6
Beilstein 9119810
Synonym 6-Chloro-3-pyridylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250mg
Melting Point 154° to 157°C

Specifications

Chemical Name or Material 6-Chloropyridine-3-boronic acid
CAS 444120-91-6
Molecular Formula C5H5BClNO2
Formula Weight 157.37
MDL Number MFCD03094998
Beilstein 9119810
Synonym 6-Chloro-3-pyridylboronic acid
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents