Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloropyridine-2-carboxylic acid, 98%

$24.86 - $201.91

Chemical Identifiers

Chemical Name or Material 6-Chloropyridine-2-carboxylic acid
CAS 4684-94-0
Molecular Formula C6H4ClNO2
Formula Weight 157.56
MDL Number MFCD00155390
Beilstein 115849
Synonym 6-Chloropicolinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19448MD

  SDS
alfa aesar™
L19448MD
250mg Each for $24.86

AAL1944806

  SDS
alfa aesar™
L1944806
5g Each for $201.91
Description & Specifications

Specifications

Formula Weight 157.56
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 6-Chloropyridine-2-carboxylic acid
Synonym 6-Chloropicolinic acid
MDL Number MFCD00155390
Quantity 250mg
Melting Point 194°C
Molecular Formula C6H4ClNO2
RTECS Number TJ7535000
Beilstein 115849
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 4684-94-0
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.