Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloropyrazine-2-carboxylic acid, 95%

Chemical Identifiers

Chemical Name or Material 6-Chloropyrazine-2-carboxylic acid
CAS 23688-89-3
Formula Weight 158.54
MDL Number MFCD00837871
Beilstein 879014
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6640403

 
alfa aesar™
H6640403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6640406

 
alfa aesar™
H6640406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66404MD

 
alfa aesar™
H66404MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.54
CAS 23688-89-3
Assay 95%
MDL Number MFCD00837871
Physical Form Solid
Quantity 1g
Beilstein 879014
Storage Note 1 -30° to -10°C
Chemical Name or Material 6-Chloropyrazine-2-carboxylic acid
Safety and Handling

-30° to -10°C