Learn More
6-Chloropurine riboside, 98%, Thermo Scientific Chemicals
$434.63 - $434.63
Chemical Identifiers
CAS | 2004-06-0 |
---|---|
Molecular Formula | C10H11ClN4O4 |
Molecular Weight (g/mol) | 286.672 |
MDL Number | MFCD00005738 |
InChI Key | XHRJGHCQQPETRH-VKZRYZQFSA-N |
Synonym | 6-chloropurine ribonucleoside |
PubChem CID | 92022056 |
IUPAC Name | (3S,4R,5S)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
SMILES | C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6461218
|
Thermo Scientific Chemicals
J6461218 |
50 g |
Each for $434.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2004-06-0 | |
286.672 | |
XHRJGHCQQPETRH-VKZRYZQFSA-N | |
92022056 | |
C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O |
C10H11ClN4O4 | |
MFCD00005738 | |
6-chloropurine ribonucleoside | |
(3S,4R,5S)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
Specifications
2004-06-0 | |
Yellow to Green | |
MFCD00005738 | |
40573 | |
XHRJGHCQQPETRH-VKZRYZQFSA-N | |
(3S,4R,5S)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
92022056 | |
98% | |
6-Chloropurine riboside |
158°C to 162°C | |
C10H11ClN4O4 | |
50 g | |
6-chloropurine ribonucleoside | |
C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O | |
286.672 | |
286.67 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-904-8
RTECSNumber : UO7520800
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only