Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloropurine riboside, 98%

Chemical Identifiers

Chemical Name or Material 6-Chloropurine riboside
CAS 5399-87-1
Formula Weight 286.67
MDL Number MFCD00005738
Beilstein 40573
Synonym 6-Chloropurine-9-β-D-ribofuranoside; 6-Chloro-9-(β-D-ribofuranosyl)purine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5963403

 
alfa aesar™
H5963403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5963406

 
alfa aesar™
H5963406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.67
Melting Point 158° to 162°C (dec.)
Chemical Name or Material 6-Chloropurine riboside
Physical Form Powder
Assay 98%
MDL Number MFCD00005738
CAS 5399-87-1
Beilstein 40573
Synonym 6-Chloropurine-9-β-D-ribofuranoside; 6-Chloro-9-(β-D-ribofuranosyl)purine
Storage Note 1 -30° to -10°C
Color White
Quantity 1g

Applications
6-Chloropurine riboside is used to study the kinetics and substrate specificity of adenosine deaminase. 6-Chloropurine riboside is benzoylated to facilitate synthesis of nucleoside derivatives such as 9-(2,3-Di-deoxy-2-fluoro-β-D-threo-pentofuranosyl)adenine. It , especially after phosphorylation to NMP, NDP or NTP, is used as a purine substrate analogue in studies with enzymes such as Inosine monophosphate dehydrogenase (IMPDH); bacteriophage T4 RNA-ligase (EC 6.5.1.3) and pancreatic fibonuclease A.

Solubility
Soluble in water. (10.6 mg/mL)

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.

Safety and Handling

-30° to -10°C