Learn More
6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals
$166.30 - $681.77
Chemical Identifiers
CAS | 4594-45-0 |
---|---|
Molecular Formula | C10H11ClN4O3 |
Molecular Weight (g/mol) | 270.673 |
MDL Number | MFCD00083282 |
InChI Key | PGEULCIODBNODW-FSDSQADBSA-N |
Synonym | 6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine |
PubChem CID | 92132234 |
IUPAC Name | (2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
SMILES | C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64871MD
|
Thermo Scientific Chemicals
J64871MD |
250 mg |
Each for $166.30
|
|
|||||
AAJ6487103
|
Thermo Scientific Chemicals
J6487103 |
1 g |
Each for $681.77
|
|
|||||
Description
6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.
Notes
Light sensitive.
Chemical Identifiers
4594-45-0 | |
270.673 | |
PGEULCIODBNODW-FSDSQADBSA-N | |
92132234 | |
C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O |
C10H11ClN4O3 | |
MFCD00083282 | |
6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine | |
(2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
Specifications
4594-45-0 | |
C10H11ClN4O3 | |
250 mg | |
6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine | |
C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O | |
270.673 | |
270.67 | |
Powder |
White | |
MFCD00083282 | |
Light sensitive | |
PGEULCIODBNODW-FSDSQADBSA-N | |
(2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
92132234 | |
97% | |
6-Chloropurine 2'-deoxyriboside |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only