Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloropiperonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 6-Chloropiperonyl chloride
CAS 23468-31-7
Molecular Formula C8H6Cl2O2
Formula Weight 205.04
MDL Number MFCD00005822
UN Number UN3261
Beilstein 150802
Synonym 5-Chloro-6-chloromethyl-1,3-benzodioxole; 2-Chloro-4,5-(methylenedioxy)benzyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0560606

 
alfa aesar™
L0560606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0560614

 
alfa aesar™
L0560614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.04
Assay 97%
Synonym 5-Chloro-6-chloromethyl-1,3-benzodioxole; 2-Chloro-4,5-(methylenedioxy)benzyl chloride
Health Hazard 1 Danger
Chemical Name or Material 6-Chloropiperonyl chloride
Molecular Formula C8H6Cl2O2
MDL Number MFCD00005822
CAS 23468-31-7
UN Number UN3261
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 62° to 66°C
Quantity 5g
Beilstein 150802

Applications
6-Chloropiperonyl chloride is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Store away from strong bases and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.