missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-chlorooxindole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC399440050

 View more versions of this product

Catalog No. AC399440050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

167.59
100.0
MFCD00209962
97.5
98%
5g
Glass bottle
6-chlorooxindole
56341-37-8
C8H6ClNO

Chemical Identifiers

56341-37-8
100.0
C8H6ClNO
MFCD00209962
97.5
98%
167.59
6-chlorooxindole

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.