missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloronicotinic acid, 99%

Manufacturer:  Alfa Aesar™ A1562006

 View more versions of this product

Catalog No. AAA1562006



Description

Description

Applications
6-Chloropyridine-3-carboxylic acid (6-chloronicotinic acid/6-CNA) has been used to study its photolytic and photocatalytic degradation. The product has been used as a media component during the isolation of 6-CNA degrading bacterial strain from imidacloprid-exposed soil samples.

Solubility
Soluble in water (2 mg/ml at 20°C), ethanol, methanol, DMSO, and slightly soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

6-Chloronicotinic acid
5g
C6H4ClNO2
115993
UAWMVMPAYRWUFX-UHFFFAOYSA-N
6-chloropyridine-3-carboxylic acid
79222
99%
~192°C (decomposition)
5326-23-8
MFCD00006241
6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid
C1=CC(=NC=C1C(=O)O)Cl
157.553
157.56

Chemical Identifiers

5326-23-8
157.553
UAWMVMPAYRWUFX-UHFFFAOYSA-N
79222
C1=CC(=NC=C1C(=O)O)Cl
C6H4ClNO2
MFCD00006241
6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid
6-chloropyridine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents