Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloronicotinamide, 98%

$67.19 - $293.82

Chemical Identifiers

Chemical Name or Material 6-Chloronicotinamide
CAS 6271-78-9
Molecular Formula C6H5ClN2O
Formula Weight 156.57
MDL Number MFCD00006242
Beilstein 116043
Synonym 6-Chloropyridine-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2192206

  SDS
alfa aesar™
B2192206
5g Each for $67.19

AAB2192214

  SDS
alfa aesar™
B2192214
25g Each for $293.82

AAB2192203

  SDS
alfa aesar™
B2192203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 6-Chloronicotinamide
CAS 6271-78-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 214° to 216°C
Quantity 5g
Assay 98%
Molecular Formula C6H5ClN2O
Formula Weight 156.57
MDL Number MFCD00006242
Beilstein 116043
Synonym 6-Chloropyridine-3-carboxamide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.