Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(Chloromethyl)uracil, 98%

$72.01 - $884.47

Chemical Identifiers

Chemical Name or Material 6-(Chloromethyl)uracil
CAS 18592-13-7
Molecular Formula C5H5ClN2O2
Formula Weight 160.56
MDL Number MFCD00010095
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2198503

  SDS
alfa aesar™
B2198503
1g Each for $72.01

AAB2198514

  SDS
alfa aesar™
B2198514
25g Each for $884.47
Description & Specifications

Specifications

Formula Weight 160.56
Chemical Name or Material 6-(Chloromethyl)uracil
TSCA TSCA
Melting Point 256° to 258°C (dec.)
Quantity 1g
Assay 98%
Molecular Formula C5H5ClN2O2
MDL Number MFCD00010095
CAS 18592-13-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.