Please login to your online account to display your discounted pricing

6-Chloromelatonin, Tocris


Melatonin agonist

Quantity 50mg

Manufacturer: r & d systems  044350

 View more versions of this product

Catalog No. 044350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 266.73
Assay Percent Range >99%
Molecular Formula C13H15ClN2O2
Chemical Name or Material 6-Chloromelatonin
Synonym N-[2-(6-Chloro-5-methoxyindol-3-yl)ethyl]acetamide
Recommended Storage Desiccate at +4°C
CAS 63762-74-3

Specifications

Chemical Name or Material 6-Chloromelatonin
Assay Percent Range >99%
Molecular Formula C13H15ClN2O2
Formula Weight 266.73
Synonym N-[2-(6-Chloro-5-methoxyindol-3-yl)ethyl]acetamide
Accessories

Desiccate at +4°C