Please login to your online account to display your discounted pricing

6-Chloromelatonin, Tocris

Melatonin agonist

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material 6-Chloromelatonin
Assay Percent Range >99%
Molecular Formula C13H15ClN2O2
Formula Weight 266.73
Synonym N-[2-(6-Chloro-5-methoxyindol-3-yl)ethyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

044310

 
r & d systems
044310
10mg Each for $113.57

044350

 
r & d systems
044350
50mg Each for $478.51
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 266.73
Chemical Name or Material 6-Chloromelatonin
Molecular Formula C13H15ClN2O2
CAS 63762-74-3
Synonym N-[2-(6-Chloro-5-methoxyindol-3-yl)ethyl]acetamide
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C