Learn More
6-Chloroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6191114
Description
6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.
Solubility
Slightly soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
6-Chloroindole-3-carboxaldehyde | |
Brown | |
MFCD06657152 | |
6-chloroindole-3-carboxaldehyde, 6-chloro indole-3-carbaldehyde, 6-chloro-3-formylindole, 6-chloro-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 6-chloro, 6-chloro-1h-indole-3-carboxaldehyde, pubchem7693, acmc-209oec, ksc494c8d, 6-chloroindole-3-carbaldehyde | |
CTNIXLBHXMSZKL-UHFFFAOYSA-N | |
6-chloro-1H-indole-3-carbaldehyde | |
12614669 | |
98% |
703-82-2 | |
C9H6ClNO | |
25 g | |
Slightly soluble in water | |
C1=CC2=C(C=C1Cl)NC=C2C=O | |
179.603 | |
179.6 | |
Powder |
Chemical Identifiers
703-82-2 | |
179.603 | |
CTNIXLBHXMSZKL-UHFFFAOYSA-N | |
12614669 | |
C1=CC2=C(C=C1Cl)NC=C2C=O |
C9H6ClNO | |
MFCD06657152 | |
6-chloroindole-3-carboxaldehyde, 6-chloro indole-3-carbaldehyde, 6-chloro-3-formylindole, 6-chloro-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 6-chloro, 6-chloro-1h-indole-3-carboxaldehyde, pubchem7693, acmc-209oec, ksc494c8d, 6-chloroindole-3-carbaldehyde | |
6-chloro-1H-indole-3-carbaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only