Please login to your online account to display your discounted pricing

6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H2Cl2N2O2S2
Formula Weight 257.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB05853CB

  SDS
fisher scientific company
SPB05853CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.12
Quantity 250mg
Assay 97%
Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
CAS 150020-64-7
Packaging Amber Glass Bottle
Molecular Formula C5H2Cl2N2O2S2
Assay Percent Range 97%