Please login to your online account to display your discounted pricing

6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB05853CB

 View more versions of this product

Catalog No. SPB05853CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 257.12
Quantity 250mg
Assay 97%
Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
Assay Percent Range 97%
Molecular Formula C5H2Cl2N2O2S2
CAS 150020-64-7
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H2Cl2N2O2S2
Formula Weight 257.12