Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid, 99%

$119.45 - $412.53

Chemical Identifiers

Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
CAS 182181-19-7
Molecular Formula C8H5ClN2O2
Formula Weight 196.59
MDL Number MFCD02186559
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5031303

  SDS
alfa aesar™
H5031303
1g Each for $412.53

AAH50313MD

  SDS
alfa aesar™
H50313MD
250mg Each for $119.45
Description & Specifications

Specifications

Formula Weight 196.59
Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Assay 99%
Molecular Formula C8H5ClN2O2
MDL Number MFCD02186559
CAS 182181-19-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 246° to 247°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.