Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chlorohexanoyl chloride, 95%

Chemical Identifiers

Chemical Name or Material 6-Chlorohexanoyl chloride
CAS 19347-73-0
Molecular Formula Cl(CH2)5COCl
Formula Weight 169.05
MDL Number MFCD00041436
UN Number UN3265
Beilstein 1852393
Synonym 6-Chlorocaproyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1111103

 
alfa aesar™
L1111103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1111106

 
alfa aesar™
L1111106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.05
Chemical Name or Material 6-Chlorohexanoyl chloride
MDL Number MFCD00041436
Assay 95%
Molecular Formula Cl(CH2)5COCl
CAS 19347-73-0
UN Number UN3265
Beilstein 1852393
Synonym 6-Chlorocaproyl chloride
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Quantity 1g
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 90°C (7mmHg)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.