missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chlorohexanoyl chloride, 95%

Manufacturer:  Alfa Aesar™ L1111106

 View more versions of this product

Catalog No. AAL1111106



Specifications

Specifications

6-Chlorohexanoyl chloride
90°C (7mmHg)
19347-73-0
MFCD00041436
1852393
WZILXAPNPKMOSA-UHFFFAOYSA-N
6-chlorohexanoyl chloride
88014
95%
>110°C (230°F)
5g
C6H10Cl2O
UN3265
hexanoyl chloride, 6-chloro, 6-chloro-caproyl chloride, acmc-1bt4p, 6-chloro-hexanoyl chloride, 6-chlorocaproic acid chloride, 6-chlorohexanoyl chloride
C(CCC(=O)Cl)CCCl
169.045
169.05

Chemical Identifiers

19347-73-0
169.045
WZILXAPNPKMOSA-UHFFFAOYSA-N
88014
C(CCC(=O)Cl)CCCl
C6H10Cl2O
MFCD00041436
hexanoyl chloride, 6-chloro, 6-chloro-caproyl chloride, acmc-1bt4p, 6-chloro-hexanoyl chloride, 6-chlorocaproic acid chloride, 6-chlorohexanoyl chloride
6-chlorohexanoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents