Please login to your online account to display your discounted pricing

6-chlorobenzo[d]isoxazol-3-ol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Chlorobenzo[d]isoxazol-3-ol
CAS 61977-29-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4ClNO2
Formula Weight 169.57
MDL Number MFCD00125033
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF01288EA

 
fisher scientific company
RF01288EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.57
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C7H4ClNO2
CAS 61977-29-5
Chemical Name or Material 6-Chlorobenzo[d]isoxazol-3-ol
Assay Percent Range 97%
MDL Number MFCD00125033
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.