missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-m-cresol, TCI America™

$22.92 - $67.22

Chemical Identifiers

CAS 615-74-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00002175
InChI Key SMFHPCZZAAMJJO-UHFFFAOYSA-N
Synonym 6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, 6-chloro-m-cresol, dsstox_cid_26558, dsstox_rid_81718
PubChem CID 12008
IUPAC Name 2-chloro-5-methylphenol
SMILES CC1=CC(=C(C=C1)Cl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C07405G View Documents TCI America
C07405G
5g Each for $22.92
 
C074025G View Documents TCI America
C074025G
25g Each for $67.22
 
Specifications

Chemical Identifiers

615-74-7
142.582
SMFHPCZZAAMJJO-UHFFFAOYSA-N
12008
CC1=CC(=C(C=C1)Cl)O
C7H7ClO
MFCD00002175
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, 6-chloro-m-cresol, dsstox_cid_26558, dsstox_rid_81718
2-chloro-5-methylphenol

Specifications

196°C
47°C
615-74-7
MFCD00002175
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, 6-chloro-m-cresol, dsstox_cid_26558, dsstox_rid_81718
CC1=CC(=C(C=C1)Cl)O
142.582
142.58
≥99.0% (GC)
Red-White
5g
C7H7ClO
3437
SMFHPCZZAAMJJO-UHFFFAOYSA-N
2-chloro-5-methylphenol
12008
Crystal-Powder at 20°C
6-Chloro-m-cresol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0900

RTECSNumber : SK4930000

TSCA : Yes