Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-alpha-phenyl-3-pyridazineacetonitrile, 97%

$342.97 - $342.97

Chemical Identifiers

Chemical Name or Material 6-Chloro-alpha-phenyl-3-pyridazineacetonitrile
CAS 73535-73-6
Molecular Formula C12H8ClN3
Formula Weight 229.67g/mol
MDL Number MFCD00140699
Synonym alpha-(6-Chloro-3-pyridazinyl)phenylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5050103

 
alfa aesar™
H50501-03
1g Each for $342.97
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 6-Chloro-alpha-phenyl-3-pyridazineacetonitrile
CAS 73535-73-6
Assay 97%
Molecular Formula C12H8ClN3
Formula Weight 229.67g/mol
MDL Number MFCD00140699
Synonym alpha-(6-Chloro-3-pyridazinyl)phenylacetonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.