missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-9-(tetrahydropyran-2-yl)purine, TCI America™

Manufacturer:  TCI America C28451G

 View more versions of this product

Catalog No. C28451G



Specifications

Specifications

6-Chloro-9-(tetrahydropyran-2-yl)purine
77°C
7306-68-5
MFCD00022821
QSTASPNCKDPSAH-UHFFFAOYSA-N
6-chloro-9-(oxan-2-yl)purine
97740
Crystal-Powder at 20°C
White-Yellow
1g
C10H11ClN4O
6-chloro-9-tetrahydro-2h-pyran-2-yl-9h-purine, 6-chloro-9-tetrahydro-2-pyranyl-purine, 6-chloro-9-tetrahydropyran-2-yl purine, 6-chloro-9-oxan-2-yl purine, 6-chloro-9-tetrahydro-2-pyranyl purine, 6-chloro-9-2-tetrahydropyranyl purine, 6-chloro-9-tetrahydro-pyran-2-yl-9h-purine, 6-chloro-9-oxan-2-yl-9h-purine, 6-chloro-9-tetrahydropyran-2-yl-9h-purine, 9h-purine, 6-chloro-9-tetrahydro-2h-pyran-2-yl
C1CCOC(C1)N2C=NC3=C2N=CN=C3Cl
238.675
238.68
≥97.0% (GC)

Chemical Identifiers

7306-68-5
238.675
QSTASPNCKDPSAH-UHFFFAOYSA-N
97740
C1CCOC(C1)N2C=NC3=C2N=CN=C3Cl
C10H11ClN4O
MFCD00022821
6-chloro-9-tetrahydro-2h-pyran-2-yl-9h-purine, 6-chloro-9-tetrahydro-2-pyranyl-purine, 6-chloro-9-tetrahydropyran-2-yl purine, 6-chloro-9-oxan-2-yl purine, 6-chloro-9-tetrahydro-2-pyranyl purine, 6-chloro-9-2-tetrahydropyranyl purine, 6-chloro-9-tetrahydro-pyran-2-yl-9h-purine, 6-chloro-9-oxan-2-yl-9h-purine, 6-chloro-9-tetrahydropyran-2-yl-9h-purine, 9h-purine, 6-chloro-9-tetrahydro-2h-pyran-2-yl
6-chloro-9-(oxan-2-yl)purine
Safety and Handling

Safety and Handling

TSCA : No

Documents