Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-9-(2-tetrahydropyranyl)purine, 99%

Chemical Identifiers

Chemical Name or Material 6-Chloro-9-(2-tetrahydropyranyl)purine
CAS 7306-68-5
Formula Weight 238.68
MDL Number MFCD00022821
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6377930

  SDS
alfa aesar™
H6377930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 149° to 151°C
Formula Weight 238.68
CAS 7306-68-5
Density 1.604g/mL
Color White to pale yellow
Refractive Index 1.7410
Chemical Name or Material 6-Chloro-9-(2-tetrahydropyranyl)purine
MDL Number MFCD00022821
Quantity 250g
Physical Form Solid
Assay 99%

Applications
6-Chloro-9-(2-tetrahydropyranyl)purine is used to prepare 6-anilino-9-(2-tetrahydropyranyl)purine by reacting with bromobenzene in the presence of potassium amide as a reagent. Further, it serves as an intermediate in organic synthesis and pharmaceuticals.

Notes
Incompatible with oxidizing agents.