Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-7-methylchromone, 98%

$142.24 - $421.89

Chemical Identifiers

Chemical Name or Material 6-Chloro-7-methylchromone
CAS 67029-84-9
Molecular Formula C10H7ClO2
Formula Weight 194.62
MDL Number MFCD00239401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699703

  SDS
alfa aesar™
H2699703
1g Each for $142.24

AAH2699706

  SDS
alfa aesar™
H2699706
5g Each for $421.89
Description & Specifications

Specifications

Formula Weight 194.62
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00239401
CAS 67029-84-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Chemical Name or Material 6-Chloro-7-methylchromone
Assay 98%
Melting Point 169° to 172°C
Molecular Formula C10H7ClO2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.