Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-7-iodo-7-deazapurine, 96%

$155.20 - $559.01

Chemical Identifiers

Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine
CAS 123148-78-7
Molecular Formula C6H3ClIN3
Formula Weight 279.47
MDL Number MFCD09263258
Synonym 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5112806

  SDS
alfa aesar™
H5112806
5g Each for $559.01

AAH5112803

  SDS
alfa aesar™
H5112803
1g Each for $155.20
Description & Specifications

Specifications

Formula Weight 279.47
MDL Number MFCD09263258
CAS 123148-78-7
Quantity 5g
Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine
Assay 96%
Molecular Formula C6H3ClIN3
Melting Point ≈180°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Synonym 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

Applications
6-Chloro-7-iodo-7-deazapurine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.