Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-5-methylpyridine-3-boronic acid, 95%

$375.19 - $861.86

Chemical Identifiers

Chemical Name or Material 6-Chloro-5-methylpyridine-3-boronic acid
CAS 1003043-40-0
Molecular Formula C6H7BClNO2
Formula Weight 171.39
MDL Number MFCD03411564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2709903

  SDS
alfa aesar™
H2709903
1g Each for $861.86

AAH27099MD

  SDS
alfa aesar™
H27099MD
250mg Each for $375.19
Description & Specifications

Specifications

Formula Weight 171.39
Quantity 1g
Chemical Name or Material 6-Chloro-5-methylpyridine-3-boronic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H7BClNO2
CAS 1003043-40-0
Melting Point 180° to 183°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
MDL Number MFCD03411564
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.