missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-5-fluorobenzimidazole, 95%

$113.69 - $1,780.15

Chemical Identifiers

CAS 175135-04-3
Molecular Formula C7H4ClFN2
Formula Weight 170.56
MDL Number MFCD00156132
Chemical Name or Material 6-Chloro-5-fluorobenzimidazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1090403 SDS Alfa Aesar™
A1090403
1g Each for $113.69
AAA1090406 SDS Alfa Aesar™
A1090406
5g Each for $439.80
AAA1090414 SDS Alfa Aesar™
A1090414
25g Each for $1,780.15
Specifications

Description

Applications
6-Chloro-5-fluorobenzimidazole is used as a pharamceutical intermediate, agrochemicals. As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment; subsequently several of the top pharmaceutical drugs contain fluorine.

Solubility
Insoluble in both water and organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

175135-04-3
170.56
6-Chloro-5-fluorobenzimidazole
C7H4ClFN2
MFCD00156132

Specifications

148° to 150°C
175135-04-3
170.56
6-Chloro-5-fluorobenzimidazole
1g
C7H4ClFN2
MFCD00156132
95%