Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-5-fluoro-2-mercaptobenzimidazole, 98%

$112.22 - $423.30

Chemical Identifiers

Chemical Name or Material 6-Chloro-5-fluoro-2-mercaptobenzimidazole
CAS 142313-30-2
Molecular Formula C7H4ClFN2S
Formula Weight 202.64
MDL Number MFCD00052490
Synonym 6-Chloro-5-fluorobenzimidazole-2-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1536703

 
alfa aesar™
A1536703
1g Each for $112.22

AAA1536706

 
alfa aesar™
A1536706
5g Each for $423.30
Description & Specifications

Specifications

Formula Weight 202.64
Molecular Formula C7H4ClFN2S
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 6-Chloro-5-fluoro-2-mercaptobenzimidazole
Assay 98%
MDL Number MFCD00052490
CAS 142313-30-2
Synonym 6-Chloro-5-fluorobenzimidazole-2-thiol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈308°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.