missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-5-fluoro-2-mercaptobenzimidazole, 98%

Manufacturer:  Alfa Aesar™ A1536706

 View more versions of this product

Catalog No. AAA1536706



Specifications

Specifications

6-Chloro-5-fluoro-2-mercaptobenzimidazole
5g
C7H4ClFN2S
6-chloro-5-fluorobenzimidazole-2-thiol, 6-chloro-5-fluoro-1h-benzo d imidazole-2-thiol, 2h-benzimidazole-2-thione,5-chloro-6-fluoro-1,3-dihydro, 5-chloro-6-fluoro-1h-1,3-benzodiazole-2-thiol, 5-chloro-6-fluoro-3h-1,3-benzodiazole-2-thiol, 5-chloro-6-fluoro-1,3-dihydro-1,3-benzodiazole-2-thione, acmc-20alzb, pubchem6710, acmc-20n1et, maybridge1_000882
C1=C2C(=CC(=C1F)Cl)NC(=S)N2
202.631
202.64
~308°C (decomposition)
142313-30-2
MFCD00052490
DQXCFZSRGLNXKW-UHFFFAOYSA-N
5-chloro-6-fluoro-1,3-dihydrobenzimidazole-2-thione
1268248
98%

Chemical Identifiers

142313-30-2
202.631
DQXCFZSRGLNXKW-UHFFFAOYSA-N
1268248
C1=C2C(=CC(=C1F)Cl)NC(=S)N2
C7H4ClFN2S
MFCD00052490
6-chloro-5-fluorobenzimidazole-2-thiol, 6-chloro-5-fluoro-1h-benzo d imidazole-2-thiol, 2h-benzimidazole-2-thione,5-chloro-6-fluoro-1,3-dihydro, 5-chloro-6-fluoro-1h-1,3-benzodiazole-2-thiol, 5-chloro-6-fluoro-3h-1,3-benzodiazole-2-thiol, 5-chloro-6-fluoro-1,3-dihydro-1,3-benzodiazole-2-thione, acmc-20alzb, pubchem6710, acmc-20n1et, maybridge1_000882
5-chloro-6-fluoro-1,3-dihydrobenzimidazole-2-thione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents