missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-4-methylpyridine-2-carboxylic acid, 95%

$47.49 - $562.88

Chemical Identifiers

CAS 324028-95-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD09834827
InChI Key IDIRHQNTRMGBPJ-UHFFFAOYSA-N
Synonym 6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid, 6-chloro-4-methyl-2-pyridinecarboxylic acid
PubChem CID 23002381
IUPAC Name 6-chloro-4-methylpyridine-2-carboxylic acid
SMILES CC1=CC(=NC(=C1)Cl)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63584MD View Documents Alfa Aesar™
H63584MD
250mg Each for $47.49
 
AAH6358403 View Documents Alfa Aesar™
H6358403
1g Each for $141.42
 
AAH6358406 View Documents Alfa Aesar™
H6358406
5g Each for $562.88
 
Specifications

Chemical Identifiers

324028-95-7
171.58
IDIRHQNTRMGBPJ-UHFFFAOYSA-N
23002381
CC1=CC(=NC(=C1)Cl)C(=O)O
C7H6ClNO2
MFCD09834827
6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid, 6-chloro-4-methyl-2-pyridinecarboxylic acid
6-chloro-4-methylpyridine-2-carboxylic acid

Specifications

White
324028-95-7
MFCD09834827
6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid, 6-chloro-4-methyl-2-pyridinecarboxylic acid
CC1=CC(=NC(=C1)Cl)C(=O)O
171.58
171.58
95%
250mg
C7H6ClNO2
UN2811
IDIRHQNTRMGBPJ-UHFFFAOYSA-N
6-chloro-4-methylpyridine-2-carboxylic acid
23002381
Solid
6-Chloro-4-methylpyridine-2-carboxylic acid
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.