Please login to your online account to display your discounted pricing

6-Chloro-4-hydroxycoumarin, 97%, ACROS Organics™

$99.66 - $99.66

Chemical Identifiers

Chemical Name or Material 6-Chloro-4-hydroxycoumarin
CAS 19484-57-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H5ClO3
Formula Weight 196.59
MDL Number MFCD00239399
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372930010

  SDS
thermo fisher scientific
AC372930010
Glass bottle 1g Each for $99.66
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 196.59
Quantity 1g
Color Slightly beige
Melting Point 270° to 275°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 6-Chloro-4-hydroxycoumarin
Assay Percent Range 97%
Molecular Formula C9H5ClO3
MDL Number MFCD00239399
CAS 19484-57-2
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.