missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-3-pyridineacetic acid, 97%, Thermo Scientific Chemicals

$133.82 - $460.19

Chemical Identifiers

CAS 39891-13-9
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD01863172
InChI Key XWVPSJPQWOVRHJ-UHFFFAOYSA-N
Synonym 2-6-chloropyridin-3-yl acetic acid, 2-chloropyridine-5-acetic acid, 6-chloro-pyridin-3-yl-acetic acid, 6-chloro-3-pyridineacetic acid, 6-chloropyridin-3-yl acetic acid, 2-6-chloro-3-pyridyl acetic acid, 3-pyridineacetic acid, 6-chloro, 2-chloropyridyl-5-acetic acid, 2-2-chloropyridin-5-yl acetic acid, 6-chloropyridine-3-acetic acid
PubChem CID 14071571
IUPAC Name 2-(6-chloropyridin-3-yl)acetic acid
SMILES C1=CC(=NC=C1CC(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2796609
View Documents Promotion Details
Thermo Scientific Chemicals
H2796609
10 g
Each for $133.82
 
AAH2796618
View Documents Promotion Details
Thermo Scientific Chemicals
H2796618
50 g
Each for $460.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39891-13-9
171.58
XWVPSJPQWOVRHJ-UHFFFAOYSA-N
14071571
C1=CC(=NC=C1CC(=O)O)Cl
C7H6ClNO2
MFCD01863172
2-6-chloropyridin-3-yl acetic acid, 2-chloropyridine-5-acetic acid, 6-chloro-pyridin-3-yl-acetic acid, 6-chloro-3-pyridineacetic acid, 6-chloropyridin-3-yl acetic acid, 2-6-chloro-3-pyridyl acetic acid, 3-pyridineacetic acid, 6-chloro, 2-chloropyridyl-5-acetic acid, 2-2-chloropyridin-5-yl acetic acid, 6-chloropyridine-3-acetic acid
2-(6-chloropyridin-3-yl)acetic acid

Specifications

39891-13-9
C7H6ClNO2
10 g
XWVPSJPQWOVRHJ-UHFFFAOYSA-N
2-(6-chloropyridin-3-yl)acetic acid
14071571
97%
172°C to 176°C
MFCD01863172
2-6-chloropyridin-3-yl acetic acid, 2-chloropyridine-5-acetic acid, 6-chloro-pyridin-3-yl-acetic acid, 6-chloro-3-pyridineacetic acid, 6-chloropyridin-3-yl acetic acid, 2-6-chloro-3-pyridyl acetic acid, 3-pyridineacetic acid, 6-chloro, 2-chloropyridyl-5-acetic acid, 2-2-chloropyridin-5-yl acetic acid, 6-chloropyridine-3-acetic acid
C1=CC(=NC=C1CC(=O)O)Cl
171.58
171.58
6-Chloro-3-pyridineacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only