Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile, 98%

$58.15 - $189.85

Chemical Identifiers

Chemical Name or Material 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile
CAS 351003-20-8
Molecular Formula C11H9ClN2O2
Formula Weight 236.65
MDL Number MFCD01834020
Synonym 6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazine-4-propionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2742303

  SDS
alfa aesar™
H2742303
1g Each for $58.15

AAH2742306

  SDS
alfa aesar™
H2742306
5g Each for $189.85
Description & Specifications

Specifications

Formula Weight 236.65
Health Hazard 1 Danger
Chemical Name or Material 6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C11H9ClN2O2
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 351003-20-8
Quantity 1g
MDL Number MFCD01834020
Melting Point 130° to 132°C
Synonym 6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazine-4-propionitrile
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.